Assay Details
Target Gene Details
Entrez Gene ID: | 91624 |
Gene Name: | nexilin F-actin binding protein |
Gene Aliases: |
CMH20, NELIN |
Location: |
Chr.1:77888515-77946329 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NEXN | NM_001172309.1 | NP_001165780.1 | ||
NM_144573.3 | NP_653174.3 | |||
XM_005271322.3 | XP_005271379.1 | |||
XM_005271323.3 | XP_005271380.1 | |||
XM_005271324.4 | XP_005271381.1 | |||
XM_005271325.3 | XP_005271382.1 | |||
XM_005271326.3 | XP_005271383.1 | |||
XM_005271327.3 | XP_005271384.1 | |||
AF114264.1 | AAD29607.1 | |||
AF176780.1 | AAQ13658.1 | |||
AK057954.1 | BAB71622.1 | |||
AK298565.1 | ||||
AK300728.1 | ||||
AK307487.1 | ||||
AY211912.1 | AAO65165.1 | |||
BC017827.1 | ||||
BC055084.1 | AAH55084.1 | |||
BC111395.1 | ||||
BC114444.1 | ||||
BC114445.1 | ||||
BX648546.1 | ||||
CR936630.1 | CAI56773.1 | |||
DQ464902.1 | ||||
S67069.1 | AAB28815.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |