Assay Details
Target Gene Details
Entrez Gene ID: | 5275 |
Gene Name: | serpin family B member 13 |
Gene Aliases: |
HSHUR7SEQ, HUR7, PI13, headpin |
Location: |
Chr.18:63587206-63599199 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SERPINB13 | NM_001307923.1 | NP_001294852.1 | ||
NM_012397.3 | NP_036529.1 | |||
XM_005266707.3 | XP_005266764.1 | |||
XM_011526029.2 | XP_011524331.1 | |||
AF169949.1 | AAD55765.1 | |||
AJ001696.2 | CAA04935.2 | |||
AJ001697.2 | CAA04936.2 | |||
AJ001698.1 | CAA04937.1 | |||
AK290323.1 | ||||
AK296797.1 | ||||
AK298049.1 | ||||
AK303867.1 | ||||
AK303979.1 | ||||
AK313486.1 | ||||
BC101821.1 | AAI01822.1 | |||
BC143402.1 | ||||
DC346089.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1066552 | Chr.18:63147299 - 64608176 on Build GRCh38 | Gain | SERPINB2 HMSD SERPINB12 SERPINB5 SERPINB3 SERPINB13 LINC00305 SERPINB8 KDSR LINC01538 VPS4B SERPINB11 SERPINB4 SERPINB10 SERPINB7 LOC284294 BCL2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |