Assay Details
Target Gene Details
Entrez Gene ID: | 652991 |
Gene Name: | SKI family transcriptional corepressor 2 |
Gene Aliases: |
CH18515, CORL2, FUSSEL18 |
Location: |
Chr.18:47206182-47249524 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SKOR2 | NM_001278063.1 | NP_001264992.1 | ||
XM_006722525.3 | XP_006722588.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3642451 | Chr.18:47089642 - 47219390 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |
nsv9629 | Chr.18:46684730 - 47231140 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)