Assay Details
Target Gene Details
Entrez Gene ID: | 55250 |
Gene Name: | elongator acetyltransferase complex subunit 2 |
Gene Aliases: |
SHINC-2, STATIP1, StIP |
Location: |
Chr.18:36129874-36177931 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ELP2 | NM_001242875.2 | NP_001229804.1 | ||
NM_001242876.2 | NP_001229805.1 | |||
NM_001242877.2 | NP_001229806.1 | |||
NM_001242878.2 | NP_001229807.1 | |||
NM_001242879.2 | NP_001229808.1 | |||
NM_001324465.1 | NP_001311394.1 | |||
NM_001324466.1 | NP_001311395.1 | |||
NM_001324467.1 | NP_001311396.1 | |||
NM_001324468.1 | NP_001311397.1 | |||
NM_018255.3 | NP_060725.1 | |||
NR_040110.2 | ||||
NR_136897.1 | ||||
NR_136898.1 | ||||
NR_137173.1 | ||||
AF332505.1 | AAK97355.1 | |||
AF403223.1 | AAQ03093.1 | |||
AK001741.1 | BAA91874.1 | |||
AK022709.1 | BAB14193.1 | |||
AK222791.1 | BAD96511.1 | |||
AK293051.1 | ||||
AK300201.1 | ||||
AK302062.1 | ||||
BC032553.1 | AAH32553.1 | |||
CR457269.1 | CAG33550.1 | |||
DC325711.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv576651 | Chr.18:36017567 - 36959615 on Build GRCh38 | Loss | LOC105372069 TPGS2 KIAA1328 SLC39A6 LOC105372071 ELP2 LOC101927809 FHOD3 MOCOS RPRD1A |
nsv576652 | Chr.18:36127201 - 36157117 on Build GRCh38 | Loss | SLC39A6 ELP2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |