Assay Details
Target Gene Details
Entrez Gene ID: | 5268 |
Gene Name: | serpin family B member 5 |
Gene Aliases: |
PI5, maspin |
Location: |
Chr.18:63476911-63505085 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SERPINB5 | NM_002639.4 | NP_002630.2 | ||
XM_006722483.2 | XP_006722546.1 | |||
AK301667.1 | ||||
AK312765.1 | ||||
CV575332.1 | ||||
U04313.1 | AAA18957.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1066552 | Chr.18:63147299 - 64608176 on Build GRCh38 | Gain | KDSR SERPINB12 LINC00305 SERPINB2 SERPINB11 BCL2 SERPINB3 LOC284294 SERPINB4 SERPINB13 VPS4B SERPINB10 SERPINB5 SERPINB7 SERPINB8 HMSD LINC01538 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |