Assay Details
Target Gene Details
Entrez Gene ID: | 23239 |
Gene Name: | PH domain and leucine rich repeat protein phosphatase 1 |
Gene Aliases: |
PHLPP, PLEKHE1, PPM3A, SCOP |
Location: |
Chr.18:62715439-62980443 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PHLPP1 | NM_194449.3 | NP_919431.2 | ||
XM_011525886.1 | XP_011524188.1 | |||
AB011178.2 | BAA25532.2 | |||
AK001924.1 | BAA91980.1 | |||
AL079409.1 | ||||
BC014927.2 | ||||
BC126277.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)