Assay Details
Target Gene Details
Entrez Gene ID: | 56853 |
Gene Name: | CUGBP, Elav-like family member 4 |
Gene Aliases: |
BRUNOL4, CELF-4 |
Location: |
Chr.18:37243040-37566052 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833621 | Chr.18:37127725 - 37302713 on Build GRCh38 | Loss | CELF4 LOC105372068 KIAA1328 |
nsv953532 | Chr.18:37246238 - 37292337 on Build GRCh38 | Deletion | CELF4 LOC105372068 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |