Assay Details
Target Gene Details
Entrez Gene ID: | 10449 |
Gene Name: | acetyl-CoA acyltransferase 2 |
Gene Aliases: |
DSAEC |
Location: |
Chr.18:49783504-49813881 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ACAA2 | NM_006111.2 | NP_006102.2 | ||
AK054673.1 | ||||
AK225457.1 | ||||
AK314462.1 | ||||
BC001918.1 | AAH01918.1 | |||
D16294.1 | BAA03800.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv516056 | Chr.18:49768512 - 49838635 on Build GRCh38 | Gain | SNHG22 ACAA2 MYO5B SCARNA17 |
nsv576863 | Chr.18:49569117 - 49938639 on Build GRCh38 | Gain | LIPG SNHG22 ACAA2 MYO5B SCARNA17 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |