Assay Details
Target Gene Details
Entrez Gene ID: | 1828 |
Gene Name: | desmoglein 1 |
Gene Aliases: |
CDHF4, DG1, DSG, EPKHE, EPKHIA, PPKS1, SPPK1 |
Location: |
Chr.18:31318089-31359246 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DSG1 | NM_001942.3 | NP_001933.2 | ||
AF097935.1 | AAC83817.1 | |||
BX476267.1 | ||||
X56654.1 | CAA39976.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |