Assay Details
Target Gene Details
Entrez Gene ID: | 59340 |
Gene Name: | histamine receptor H4 |
Gene Aliases: |
AXOR35, BG26, GPCR105, GPRv53, H4, H4R, HH4R |
Location: |
Chr.18:24460568-24487427 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HRH4 | NM_001143828.1 | NP_001137300.1 | ||
NM_001160166.1 | NP_001153638.1 | |||
NM_021624.3 | NP_067637.2 | |||
XM_011526133.1 | XP_011524435.1 | |||
AB044934.1 | BAB13698.1 | |||
AB045370.1 | BAB20091.1 | |||
AF307973.1 | AAG32052.1 | |||
AF312230.1 | AAK12081.1 | |||
AF325356.1 | AAL01684.1 | |||
AF329449.1 | AAK43542.1 | |||
AJ298292.1 | CAC83493.1 | |||
AY008280.1 | AAL09297.1 | |||
AY136745.1 | AAN01271.1 | |||
BC069136.1 | AAH69136.1 | |||
BC112348.1 | AAI12349.1 | |||
DQ835186.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv521369 | Chr.18:24468583 - 24479130 on Build GRCh38 | Gain | HRH4 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |