Assay Details
Target Gene Details
Entrez Gene ID: | 147429 |
Gene Name: | AQP4 antisense RNA 1 |
Gene Aliases: |
C18orf16, CHST9-AS1 |
Location: |
Chr.18:26865308-26935946 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AQP4-AS1 | NR_026908.1 | |||
AK055069.1 | BAB70850.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833600 | Chr.18:26893930 - 27101055 on Build GRCh38 | Gain | CHST9 AQP4-AS1 |
esv2761995 | Chr.18:26634322 - 26922606 on Build GRCh38 | Loss | KCTD1 CHST9 AQP4 PCAT18 AQP4-AS1 |