Assay Details
Target Gene Details
Entrez Gene ID: | 2774 |
Gene Name: | G protein subunit alpha L |
Gene Aliases: |
DYT25 |
Location: |
Chr.18:11688956-11885685 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GNAL | NM_182978.3 | NP_892023.1 | ||
XM_006722324.2 | XP_006722387.1 | |||
AK090868.1 | BAC03535.1 | |||
AK295088.1 | ||||
AK307260.1 | ||||
BC050021.1 | AAH50021.1 | |||
BP230966.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1057995 | Chr.18:10626172 - 11923368 on Build GRCh38 | Gain | GNAL LOC101927410 MIR7153 MIR6788 PIEZO2 LINC01255 SLC35G4 MPPE1 CHMP1B |
nsv518207 | Chr.18:11670846 - 11695677 on Build GRCh38 | Loss | GNAL |
nsv516128 | Chr.18:11683432 - 11695677 on Build GRCh38 | Loss | GNAL |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |