Assay Details
Target Gene Details
Entrez Gene ID: | 58488 |
Gene Name: | phosphatidylcholine transfer protein |
Gene Aliases: |
PC-TP, STARD2 |
Location: |
Chr.17:55750979-55845001 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PCTP | NM_001102402.2 | NP_001095872.1 | ||
NM_021213.3 | NP_067036.2 | |||
XM_005257549.1 | XP_005257606.1 | |||
XM_005257550.3 | XP_005257607.1 | |||
XM_017024913.1 | XP_016880402.1 | |||
AF087857.1 | AAP97167.1 | |||
AF114430.1 | AAF08345.1 | |||
AF151638.1 | AAF02536.1 | |||
AK024667.1 | ||||
AK058120.1 | ||||
AK314509.1 | ||||
BC005112.1 | AAH05112.1 | |||
BC012084.1 | AAH12084.1 | |||
BM153142.1 | ||||
BM549250.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1076450 | Chr.17:55754638 - 55761639 on Build GRCh38 | Duplication | PCTP |
nsv2100 | Chr.17:55415019 - 56251155 on Build GRCh38 | Deletion | ANKFN1 TMEM100 PCTP MMD |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |