Assay Details
Target Gene Details
Entrez Gene ID: | 100529207 |
Gene Name: | RAD51L3-RFFL readthrough |
Gene Aliases: |
- |
Location: |
Chr.17:35009112-35121522 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RAD51L3-RFFL | NR_037714.1 | |||
AK097811.1 |
Target Gene Details
Entrez Gene ID: | 117584 |
Gene Name: | ring finger and FYVE-like domain containing E3 ubiquitin protein ligase |
Gene Aliases: |
CARP-2, CARP2, FRING, RIFIFYLIN, RNF189, RNF34L |
Location: |
Chr.17:35009112-35089329 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RFFL | NM_001017368.1 | NP_001017368.1 | ||
NR_037713.1 | ||||
AF434816.1 | AAL30771.1 | |||
AK093112.1 | BAC04059.1 | |||
AK308689.1 | ||||
AY098935.1 | AAM29181.1 | |||
BC015681.2 | AAH15681.2 | |||
BC028424.1 | AAH28424.1 | |||
CR933651.1 | CAI45952.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv2035 | Chr.17:35002602 - 35047830 on Build GRCh38 | Deletion | RFFL LIG3 RAD51L3-RFFL |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |