Assay Details
Target Gene Details
Entrez Gene ID: | 8844 |
Gene Name: | kinase suppressor of ras 1 |
Gene Aliases: |
KSR, RSU2 |
Location: |
Chr.17:27456441-27626438 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KSR1 | XM_006722154.3 | XP_006722217.1 | ||
XM_011525429.2 | XP_011523731.1 | |||
XM_011525430.2 | XP_011523732.1 | |||
XM_011525431.2 | XP_011523733.1 | |||
XM_017025267.1 | XP_016880756.1 | |||
XM_017025268.1 | XP_016880757.1 | |||
XM_017025269.1 | XP_016880758.1 | |||
XM_017025279.1 | XP_016880768.1 | |||
XM_017025280.1 | XP_016880769.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv540e199 | Chr.17:26964108 - 27464626 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)