Assay Details
Target Gene Details
Entrez Gene ID: | 11325 |
Gene Name: | DEAD-box helicase 42 |
Gene Aliases: |
DDX42P, RHELP, RNAHP, SF3B8, SF3b125 |
Location: |
Chr.17:63773856-63819317 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DDX42 | NM_007372.3 | NP_031398.2 | ||
NM_203499.2 | NP_987095.1 | |||
XM_006721657.1 | XP_006721720.1 | |||
XM_017024111.1 | XP_016879600.1 | |||
AL527653.3 | ||||
AL545851.3 | ||||
BC093081.1 | AAH93081.1 | |||
DA440475.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833510 | Chr.17:63767293 - 63927790 on Build GRCh38 | Loss | SMARCD2 LOC107987248 GH1 PSMC5 CSHL1 CSH2 DDX42 CSH1 GH2 FTSJ3 CCDC47 TCAM1P |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |