Assay Details
Target Gene Details
Entrez Gene ID: | 105369205 |
Gene Name: | uncharacterized LOC105369205 |
Gene Aliases: |
- |
Location: |
Chr.17:20921016-21001059 on Build GRCh38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC105369205 |
Target Gene Details
Entrez Gene ID: | 339260 |
Gene Name: | uncharacterized LOC339260 |
Gene Aliases: |
- |
Location: |
Chr.17:20938520-20982357 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC339260 | NR_123730.1 | |||
DB066214.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1064365 | Chr.17:20898426 - 20987403 on Build GRCh38 | Gain | LOC339260 CCDC144NL-AS1 LOC105369205 |
esv2751619 | Chr.17:20559595 - 21010843 on Build GRCh38 | Gain | LOC339260 CCDC144NL CDRT15L2 LOC100287072 CCDC144NL-AS1 MEIS3P2 LOC105369205 USP22 ABHD17AP6 |
nsv1059146 | Chr.17:20849194 - 22546654 on Build GRCh38 | Gain | DHRS7B MTRNR2L1 FLJ36000 CCDC144NL UBBP4 CCDC144NL-AS1 LOC105369205 KCNJ18 C17orf51 NATD1 MAP2K3 KCNJ12 LOC339260 LINC01563 TMEM11 LOC100996792 FAM27E5 USP22 |
esv3640224 | Chr.17:20957522 - 20982745 on Build GRCh38 | Loss | LOC339260 LOC105369205 |
nsv2006 | Chr.17:20943450 - 20989417 on Build GRCh38 | Deletion | LOC339260 LOC105369205 |