Assay Details
Target Gene Details
Entrez Gene ID: | 54890 |
Gene Name: | alkB homolog 5, RNA demethylase |
Gene Aliases: |
ABH5, OFOXD, OFOXD1 |
Location: |
Chr.17:18183553-18209954 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ALKBH5 | NM_017758.3 | NP_060228.3 | ||
AK000315.1 | BAA91078.1 | |||
AK301107.1 | ||||
AK311161.1 | ||||
AK311511.1 | ||||
BC062339.1 | AAH62339.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv510702 | Chr.17:18134362 - 18213487 on Build GRCh38 | Deletion | ALKBH5 MYO15A |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |