Assay Details
Target Gene Details
Entrez Gene ID: | 51082 |
Gene Name: | RNA polymerase I subunit D |
Gene Aliases: |
AC19, POLR1C, RPA16, RPA9, RPAC2, RPC16, RPO1-3, TCS2 |
Location: |
Chr.13:27620743-27667422 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
POLR1D | NM_001206559.1 | NP_001193488.1 | ||
NM_152705.2 | NP_689918.1 | |||
XM_005266414.2 | XP_005266471.1 | |||
XM_017020622.1 | XP_016876111.1 | |||
XM_017020623.1 | XP_016876112.1 | |||
AK097973.1 | ||||
AK307356.1 | ||||
AK311833.1 | ||||
BC006972.1 | AAH06972.1 | |||
BC015319.2 | AAH15319.1 | |||
BC018528.1 | AAH18528.1 | |||
DB455307.1 | ||||
DB459107.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |