Assay Details
Target Gene Details
Entrez Gene ID: | 2259 |
Gene Name: | fibroblast growth factor 14 |
Gene Aliases: |
FGF-14, FHF-4, FHF4, SCA27 |
Location: |
Chr.13:101720855-102402428 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv832702 | Chr.13:101621711 - 101795212 on Build GRCh38 | Gain |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)