Assay Details
Target Gene Details
Entrez Gene ID: | 160897 |
Gene Name: | G protein-coupled receptor 180 |
Gene Aliases: |
ITR |
Location: |
Chr.13:94601850-94634645 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GPR180 | NM_180989.5 | NP_851320.1 | ||
AK126794.1 | BAC86697.1 | |||
AK289850.1 | ||||
BC052243.1 | AAH52243.1 | |||
CD110068.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv832686 | Chr.13:94554488 - 94748738 on Build GRCh38 | Gain | GPR180 SOX21 TGDS SOX21-AS1 LOC101927248 |
nsv832685 | Chr.13:94527422 - 94627712 on Build GRCh38 | Gain | GPR180 TGDS DCT |
esv34004 | Chr.13:94254283 - 94740390 on Build GRCh38 | Loss | GPR180 GPC6 SOX21 TGDS SOX21-AS1 DCT LOC101927248 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |