Assay Details
Target Gene Details
Entrez Gene ID: | 160851 |
Gene Name: | diacylglycerol kinase eta |
Gene Aliases: |
DGKeta |
Location: |
Chr.13:42040036-42256584 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DGKH | NM_001204504.2 | NP_001191433.1 | ||
NM_152910.5 | NP_690874.2 | |||
NM_178009.4 | NP_821077.1 | |||
AB078967.1 | BAC66960.1 | |||
AB078968.1 | BAC66961.1 | |||
AK289546.1 | ||||
DR002010.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv832588 | Chr.13:41876984 - 42053116 on Build GRCh38 | Gain | VWA8-AS1 VWA8 DGKH |
nsv1016 | Chr.13:42041861 - 42072289 on Build GRCh38 | Insertion | DGKH |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |