Assay Details
Target Gene Details
Entrez Gene ID: | 2842 |
Gene Name: | G protein-coupled receptor 19 |
Gene Aliases: |
- |
Location: |
Chr.12:12659691-12716305 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GPR19 | NM_006143.2 | NP_006134.1 | ||
XM_011520623.2 | XP_011518925.1 | |||
XM_011520624.2 | XP_011518926.1 | |||
XM_011520625.2 | XP_011518927.1 | |||
XM_017019212.1 | XP_016874701.1 | |||
XM_017019213.1 | XP_016874702.1 | |||
XM_017019214.1 | XP_016874703.1 | |||
XM_017019215.1 | XP_016874704.1 | |||
XM_017019216.1 | XP_016874705.1 | |||
AK074729.1 | ||||
DB022290.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv89n21 | Chr.12:12657720 - 12677103 on Build GRCh38 | Gain |
![]() |
esv2745567 | Chr.12:12246086 - 13092998 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv509456 | Chr.12:12645094 - 12681838 on Build GRCh38 | Insertion |
![]() ![]() |
nsv1046355 | Chr.12:11501836 - 12890185 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)