Assay Details
Target Gene Details
Entrez Gene ID: | 171017 |
Gene Name: | zinc finger protein 384 |
Gene Aliases: |
CAGH1, CAGH1A, CIZ, ERDA2, NMP4, NP, TNRC1 |
Location: |
Chr.12:6666477-6689572 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv832322 | Chr.12:6575884 - 6765914 on Build GRCh38 | Loss | COPS7A SCARNA11 ZNF384 MLF2 PIANP ING4 LPAR5 ACRBP CHD4 |
nsv832323 | Chr.12:6662233 - 6856559 on Build GRCh38 | Loss | PTMS ZNF384 GPR162 GNB3 USP5 COPS7A MLF2 PIANP P3H3 ING4 CDCA3 LAG3 CD4 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |