Assay Details
Target Gene Details
Entrez Gene ID: | 5288 |
Gene Name: | phosphatidylinositol-4-phosphate 3-kinase catalytic subunit type 2 gamma |
Gene Aliases: |
PI3K-C2-gamma, PI3K-C2GAMMA |
Location: |
Chr.12:18242946-18716861 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PIK3C2G | NM_001288772.1 | NP_001275701.1 | ||
NM_001288774.1 | NP_001275703.1 | |||
NM_004570.5 | NP_004561.3 | |||
XM_011520696.2 | XP_011518998.1 | |||
XM_011520697.2 | XP_011518999.1 | |||
XM_011520700.2 | XP_011519002.1 | |||
XM_011520701.2 | XP_011519003.1 | |||
XM_017019470.1 | XP_016874959.1 | |||
XM_017019471.1 | XP_016874960.1 | |||
XM_017019472.1 | XP_016874961.1 | |||
XM_017019473.1 | XP_016874962.1 | |||
XM_017019474.1 | XP_016874963.1 | |||
XM_017019475.1 | XP_016874964.1 | |||
XM_017019476.1 | XP_016874965.1 | |||
XM_017019478.1 | XP_016874967.1 | |||
AJ000008.1 | CAA03853.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3892152 | Chr.12:17539788 - 18350960 on Build GRCh38 | Gain |
![]() ![]() ![]() |
nsv557667 | Chr.12:18123815 - 18628015 on Build GRCh38 | Gain |
![]() |
nsv832344 | Chr.12:18138275 - 18353053 on Build GRCh38 | Loss |
![]() |
dgv2395n54 | Chr.12:18223260 - 18350960 on Build GRCh38 | Loss |
![]() |
nsv832343 | Chr.12:18126546 - 18307230 on Build GRCh38 | Gain |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)