Assay Details
Target Gene Details
Entrez Gene ID: | 89910 |
Gene Name: | ubiquitin protein ligase E3B |
Gene Aliases: |
BPIDS, KOS |
Location: |
Chr.12:109477410-109543628 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
UBE3B | NM_001270449.1 | NP_001257378.1 | ||
NM_001270450.1 | NP_001257379.1 | |||
NM_001270451.1 | NP_001257380.1 | |||
NM_130466.3 | NP_569733.2 | |||
NM_183415.2 | NP_904324.1 | |||
XM_005253987.2 | XP_005254044.1 | |||
XM_006719681.3 | XP_006719744.1 | |||
XM_006719682.2 | XP_006719745.1 | |||
XM_011538959.2 | XP_011537261.1 | |||
XM_011538961.1 | XP_011537263.1 | |||
XM_017020195.1 | XP_016875684.1 | |||
XM_017020196.1 | XP_016875685.1 | |||
AF251046.2 | AAK28419.2 | |||
AK307385.1 | ||||
AL096740.1 | CAH56410.1 | |||
BC032301.1 | ||||
BC051266.1 | AAH51266.1 | |||
BC068221.1 | AAH68221.1 | |||
BC108705.1 | AAI08706.1 | |||
BC141880.1 | ||||
BC150637.1 | ||||
BX537403.1 | CAD97645.1 | |||
DA771082.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |