Assay Details
Target Gene Details
Entrez Gene ID: | 6996 |
Gene Name: | thymine DNA glycosylase |
Gene Aliases: |
hTDG |
Location: |
Chr.12:103965815-103988878 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TDG | NM_003211.4 | NP_003202.3 | ||
XM_005269125.1 | XP_005269182.1 | |||
XM_011538714.1 | XP_011537016.1 | |||
XM_011538715.1 | XP_011537017.1 | |||
AK299832.1 | ||||
AK303631.1 | ||||
AK309792.1 | ||||
AK311673.1 | ||||
AK313234.1 | ||||
AL710869.1 | ||||
BC019925.1 | ||||
BC037557.1 | AAH37557.1 | |||
BC104477.1 | AAI04478.1 | |||
CB047650.1 | ||||
DB162106.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv560039 | Chr.12:103783890 - 104061562 on Build GRCh38 | Gain | GLT8D2 HSP90B1 MIR3652 NT5DC3 C12orf73 TTC41P TDG |
esv2746302 | Chr.12:103966022 - 103979831 on Build GRCh38 | Deletion | TDG |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |