Assay Details
Target Gene Details
Entrez Gene ID: | 25834 |
Gene Name: | MGAT4 family member C |
Gene Aliases: |
GNTIVH, HGNT-IV-H |
Location: |
Chr.12:85972281-86838999 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MGAT4C | NM_013244.3 | NP_037376.2 | ||
XM_005268776.4 | XP_005268833.1 | |||
XM_005268779.3 | XP_005268836.1 | |||
XM_011538150.2 | XP_011536452.1 | |||
XM_011538151.2 | XP_011536453.1 | |||
XM_011538156.2 | XP_011536458.1 | |||
XM_011538157.2 | XP_011536459.1 | |||
XM_017019143.1 | XP_016874632.1 | |||
XM_017019144.1 | XP_016874633.1 | |||
XM_017019145.1 | XP_016874634.1 | |||
XM_017019146.1 | XP_016874635.1 | |||
XM_017019147.1 | XP_016874636.1 | |||
XM_017019148.1 | XP_016874637.1 | |||
XM_017019149.1 | XP_016874638.1 | |||
XM_017019150.1 | XP_016874639.1 | |||
AK127727.1 | ||||
AK299253.1 | ||||
BC026068.1 | AAH26068.1 | |||
BC064141.1 | AAH64141.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv559667 | Chr.12:85924226 - 86022434 on Build GRCh38 | Loss | MGAT4C |
nsv1049179 | Chr.12:85662598 - 86405410 on Build GRCh38 | Gain | RASSF9 NTS MGAT4C |
esv3892241 | Chr.12:85305039 - 88102633 on Build GRCh38 | Gain | C12orf29 C12orf50 RASSF9 LOC105369879 MKRN9P NTS CEP290 MGAT4C |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |