Assay Details
Target Gene Details
Entrez Gene ID: | 100287944 |
Gene Name: | uncharacterized LOC100287944 |
Gene Aliases: |
- |
Location: |
Chr.12:106496410-106774831 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC100287944 | NR_040246.1 | |||
DA250223.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv832510 | Chr.12:106393952 - 106557362 on Build GRCh38 | Loss | POLR3B LOC100287944 |