Assay Details
Target Gene Details
Entrez Gene ID: | 81539 |
Gene Name: | solute carrier family 38 member 1 |
Gene Aliases: |
ATA1, NAT2, SAT1, SNAT1 |
Location: |
Chr.12:46183055-46269425 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC38A1 | NM_001077484.1 | NP_001070952.1 | ||
NM_001278387.1 | NP_001265316.1 | |||
NM_001278388.1 | NP_001265317.1 | |||
NM_001278389.1 | NP_001265318.1 | |||
NM_001278390.1 | NP_001265319.1 | |||
NM_030674.3 | NP_109599.3 | |||
XM_011538784.2 | XP_011537086.1 | |||
XM_011538786.2 | XP_011537088.1 | |||
XM_011538787.2 | XP_011537089.1 | |||
XM_017019990.1 | XP_016875479.1 | |||
AF247166.1 | AAG44546.1 | |||
AF271070.1 | AAG39354.1 | |||
AK027825.1 | BAB55394.1 | |||
AK074797.1 | BAC11215.1 | |||
AK074851.1 | ||||
AK074949.1 | BAC11310.1 | |||
AK225639.1 | ||||
AK225778.1 | ||||
AK308129.1 | ||||
AK309410.1 | ||||
AK313731.1 | ||||
BC010620.1 | AAH10620.1 | |||
BX647547.1 | ||||
BX648495.1 | ||||
BX648676.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)