Assay Details
Target Gene Details
Entrez Gene ID: | 57494 |
Gene Name: | ribosomal modification protein rimK like family member B |
Gene Aliases: |
FAM80B, NAAGS, NAAGS-I |
Location: |
Chr.12:8681606-8783098 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RIMKLB | NR_123740.1 | |||
XM_017019684.1 | XP_016875173.1 | |||
XM_017019689.1 | XP_016875178.1 | |||
XM_017019697.1 | XP_016875186.1 | |||
DB265022.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1051530 | Chr.12:8442286 - 9951690 on Build GRCh38 | Gain | CLEC12A RIMKLB CLEC2D AICDA LOC101930452 LOC101928030 LOC400002 DDX12P CLEC4E CLEC6A KLRF1 LOC105369728 MIR1244-3 CLEC2B PHC1 LOC728715 PZP LOC105369649 KLRF2 CD69 A2M M6PR LOC374443 A2M-AS1 LOC642846 CLECL1 KLRG1 A2ML1 LINC00612 CLEC2A A2MP1 OVOS CLEC4D MFAP5 LOC105376678 LINC00987 LOC100506159 KLRB1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |