Assay Details
Target Gene Details
Entrez Gene ID: | 5031 |
Gene Name: | pyrimidinergic receptor P2Y6 |
Gene Aliases: |
P2Y6 |
Location: |
Chr.11:73264503-73298625 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
P2RY6 | NM_001277204.1 | NP_001264133.1 | ||
NM_001277205.1 | NP_001264134.1 | |||
NM_001277206.1 | NP_001264135.1 | |||
NM_001277207.1 | NP_001264136.1 | |||
NM_001277208.1 | NP_001264137.1 | |||
NM_176796.2 | NP_789766.1 | |||
NM_176797.2 | NP_789767.1 | |||
NM_176798.2 | NP_789768.1 | |||
XM_011545076.2 | XP_011543378.1 | |||
XM_011545077.2 | XP_011543379.1 | |||
XM_011545079.2 | XP_011543381.1 | |||
AF007891.1 | AAB80712.1 | |||
AF007892.1 | AAB80713.1 | |||
AF007893.1 | ||||
AF218005.1 | AAG17247.1 | |||
AL520218.3 | ||||
AL525099.3 | ||||
AX775903.1 | CAE11651.1 | |||
BC000571.2 | AAH00571.1 | |||
BC009391.2 | AAH09391.1 | |||
BQ718871.1 | ||||
BX337204.2 | ||||
BX381745.2 | ||||
BX397649.2 | ||||
DR004482.1 | ||||
DR006998.1 | ||||
U52464.1 | AAB03572.1 | |||
X97058.1 | CAA65770.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv387 | Chr.11:73253689 - 73285467 on Build GRCh38 | Insertion | P2RY6 |
nsv832208 | Chr.11:73165139 - 73379958 on Build GRCh38 | Gain | ARHGEF17 RELT P2RY2 P2RY6 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |