Assay Details
Target Gene Details
Entrez Gene ID: | 5029 |
Gene Name: | purinergic receptor P2Y2 |
Gene Aliases: |
HP2U, P2RU1, P2U, P2U1, P2UR, P2Y2, P2Y2R |
Location: |
Chr.11:73200416-73246743 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
P2RY2 | XM_017017839.1 | XP_016873328.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2669809 | Chr.11:73203051 - 73203990 on Build GRCh38 | Deletion | P2RY2 |
esv3626811 | Chr.11:73203052 - 73203993 on Build GRCh38 | Loss | P2RY2 |
nsv832208 | Chr.11:73165139 - 73379958 on Build GRCh38 | Gain | RELT ARHGEF17 P2RY2 P2RY6 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |