Assay Details
Target Gene Details
Entrez Gene ID: | 1656 |
Gene Name: | DEAD-box helicase 6 |
Gene Aliases: |
HLR2, P54, RCK |
Location: |
Chr.11:118747763-118791263 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DDX6 | NM_001257191.2 | NP_001244120.1 | ||
NM_004397.5 | NP_004388.2 | |||
XM_005271417.3 | XP_005271474.1 | |||
XM_011542644.1 | XP_011540946.1 | |||
XM_011542645.1 | XP_011540947.1 | |||
XM_017017251.1 | XP_016872740.1 | |||
AK313244.1 | ||||
BC039826.1 | AAH39826.1 | |||
BC065007.1 | AAH65007.1 | |||
D17532.1 | BAA04482.1 | |||
Z11685.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv832283 | Chr.11:118566790 - 118786484 on Build GRCh38 | Loss | IFT46 ARCN1 TREH MIR6716 DDX6 PHLDB1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |