Assay Details
Target Gene Details
Entrez Gene ID: | 26297 |
Gene Name: | secretion regulating guanine nucleotide exchange factor |
Gene Aliases: |
DELGEF, Gnefr |
Location: |
Chr.11:17788048-18013162 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SERGEF | NM_012139.3 | NP_036271.1 | ||
NR_104040.1 | ||||
NR_104041.1 | ||||
AF086056.1 | ||||
AJ243950.1 | CAB60832.1 | |||
AJ243951.1 | CAB60833.1 | |||
AK292286.1 | ||||
BC065375.1 | AAH65375.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv553567 | Chr.11:16890704 - 17946708 on Build GRCh38 | Gain | SNORD14B MYOD1 USH1C LOC102723330 LOC105376575 SNORD14A KCNJ11 OTOG SERGEF NUCB2 PLEKHA7 ABCC8 PIK3C2A OR7E14P RPS13 NCR3LG1 KCNC1 |
nsv509390 | Chr.11:17726193 - 17855296 on Build GRCh38 | Insertion | SERGEF KCNC1 |
nsv832078 | Chr.11:17758956 - 17931974 on Build GRCh38 | Loss | SERGEF KCNC1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |