Assay Details
Target Gene Details
Entrez Gene ID: | 57689 |
Gene Name: | leucine rich repeat containing 4C |
Gene Aliases: |
NGL-1, NGL1 |
Location: |
Chr.11:40107066-41460212 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LRRC4C | XM_011520238.2 | 8 | 9879 | XP_011518540.1 |
XM_011520239.2 | 9 | 9945 | XP_011518541.1 | |
XM_011520240.2 | 7 | 9799 | XP_011518542.1 | |
XM_011520241.2 | 4 | 10514 | XP_011518543.1 | |
XM_011520242.2 | 4 | 10500 | XP_011518544.1 | |
XM_011520243.2 | 7 | 9826 | XP_011518545.1 | |
XM_011520244.2 | 6 | 9746 | XP_011518546.1 | |
XM_017018070.1 | 8 | 9875 | XP_016873559.1 | |
XM_017018071.1 | 8 | 9865 | XP_016873560.1 | |
XM_017018072.1 | 8 | 9728 | XP_016873561.1 | |
XM_017018073.1 | 6 | 9587 | XP_016873562.1 | |
XM_017018074.1 | 4 | 10495 | XP_016873563.1 | |
XM_017018075.1 | 5 | 9534 | XP_016873564.1 | |
XM_017018076.1 | 3 | 10461 | XP_016873565.1 | |
XM_017018077.1 | 6 | 9614 | XP_016873566.1 | |
XM_017018078.1 | 5 | 10580 | XP_016873567.1 | |
XM_017018079.1 | 6 | 9742 | XP_016873568.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv65n21 | Chr.11:40049473 - 40249892 on Build GRCh38 | Gain | LRRC4C |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |