Assay Details
Target Gene Details
Entrez Gene ID: | 1737 |
Gene Name: | dihydrolipoamide S-acetyltransferase |
Gene Aliases: |
DLTA, PDC-E2, PDCE2 |
Location: |
Chr.11:112024814-112064278 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DLAT | NM_001931.4 | NP_001922.2 | ||
XM_011542647.2 | XP_011540949.1 | |||
AK057299.1 | ||||
AK223596.1 | BAD97316.1 | |||
AK296274.1 | ||||
AK297103.1 | ||||
AK299562.1 | ||||
AK303930.1 | ||||
AL832703.1 | ||||
BC039084.1 | AAH39084.1 | |||
Y00978.1 | CAA68787.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2745077 | Chr.11:111949712 - 112188677 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)