Assay Details
Target Gene Details
Entrez Gene ID: | 80310 |
Gene Name: | platelet derived growth factor D |
Gene Aliases: |
IEGF, MSTP036, SCDGF-B, SCDGFB |
Location: |
Chr.11:103907186-104164299 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PDGFD | NM_025208.4 | NP_079484.1 | ||
NM_033135.3 | NP_149126.1 | |||
AB033832.1 | BAB18903.1 | |||
AF113216.1 | AAG39287.1 | |||
AF335584.1 | AAK38840.1 | |||
AF336376.1 | AAK56136.1 | |||
AK292801.1 | ||||
AY027517.1 | AAK20081.1 | |||
AY027518.1 | AAK20082.1 | |||
AY359116.1 | AAQ89474.1 | |||
BC030645.2 | AAH30645.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2750977 | Chr.11:103211829 - 104054062 on Build GRCh38 | Gain | PDGFD DYNC2H1 DDI1 MIR4693 |
esv2760322 | Chr.11:103889357 - 104503413 on Build GRCh38 | Loss | PDGFD DDI1 MIR7641-1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |