Assay Details
Target Gene Details
Entrez Gene ID: | 9440 |
Gene Name: | mediator complex subunit 17 |
Gene Aliases: |
CRSP6, CRSP77, DRIP80, TRAP80 |
Location: |
Chr.11:93784239-93813330 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MED17 | NM_004268.4 | NP_004259.3 | ||
XM_011543068.2 | XP_011541370.1 | |||
AF105421.1 | AAD30856.1 | |||
AF117657.2 | AAD22031.2 | |||
AK001674.1 | BAA91827.1 | |||
AK023209.1 | ||||
AK299702.1 | ||||
BC021101.1 | AAH21101.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv832232 | Chr.11:93668148 - 93816272 on Build GRCh38 | Gain | C11orf54 SCARNA9 SNORA25 SNORA32 MIR1304 SNORA8 TAF1D SNORD6 SNORA40 CEP295 SNORA18 MED17 SNORA1 SNORD5 |
esv3627294 | Chr.11:93485306 - 93944317 on Build GRCh38 | Gain | C11orf54 SCARNA9 SNORA25 VSTM5 SNORA32 MIR1304 SMCO4 SNORA8 TAF1D SNORD6 SNORA40 CEP295 SNORA18 MED17 SNORA1 SNORD5 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |