Assay Details
Target Gene Details
Entrez Gene ID: | 79827 |
Gene Name: | CXADR like membrane protein |
Gene Aliases: |
ACAM, ASAM, CSBM, CSBS |
Location: |
Chr.11:123072006-123195305 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CLMP | NM_024769.3 | NP_079045.1 | ||
XM_011542998.2 | XP_011541300.1 | |||
AK026068.1 | BAB15347.1 | |||
AK304843.1 | ||||
AY326422.1 | AAP88386.1 | |||
AY358340.1 | AAQ88706.1 | |||
BC009371.2 | AAH09371.1 | |||
BE272517.1 | ||||
BG723094.1 | ||||
CA395814.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |