Assay Details
Target Gene Details
Entrez Gene ID: | 83700 |
Gene Name: | junctional adhesion molecule 3 |
Gene Aliases: |
JAM-2, JAM-3, JAM-C, JAMC |
Location: |
Chr.11:134068925-134151757 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
JAM3 | NM_001205329.1 | NP_001192258.1 | ||
NM_032801.4 | NP_116190.3 | |||
AF356518.1 | AAK27221.1 | |||
AF448478.1 | AAM20925.1 | |||
AJ344431.1 | CAC69845.1 | |||
AJ416101.1 | CAC94776.1 | |||
AK074769.1 | BAC11195.1 | |||
AK075309.1 | BAC11538.1 | |||
AK125071.1 | ||||
AK225666.1 | ||||
AY358335.1 | AAQ88701.1 | |||
BC010690.1 | AAH10690.1 | |||
BC012147.1 | AAH12147.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3628161 | Chr.11:134108317 - 134113467 on Build GRCh38 | Loss | JAM3 |
esv3892114 | Chr.11:134054794 - 134328278 on Build GRCh38 | Gain | VPS26B ACAD8 THYN1 PTP4A2P2 GLB1L3 JAM3 NCAPD3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |