Assay Details
Target Gene Details
Entrez Gene ID: | 38 |
Gene Name: | acetyl-CoA acetyltransferase 1 |
Gene Aliases: |
ACAT, MAT, T2, THIL |
Location: |
Chr.11:108121531-108148168 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ACAT1 | NM_000019.3 | NP_000010.1 | ||
XM_006718835.3 | XP_006718898.1 | |||
XM_017017681.1 | XP_016873170.1 | |||
XM_017017682.1 | XP_016873171.1 | |||
XM_017017683.1 | XP_016873172.1 | |||
AK312574.1 | ||||
BC010942.1 | AAH10942.1 | |||
D90228.1 | BAA14278.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv1278n100 | Chr.11:108079845 - 108178945 on Build GRCh38 | Gain |
![]() ![]() ![]() |
nsv480 | Chr.11:108112138 - 108151779 on Build GRCh38 | Insertion |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)