Assay Details
Target Gene Details
Entrez Gene ID: | 51074 |
Gene Name: | APAF1 interacting protein |
Gene Aliases: |
APIP2, CGI-29, CGI29, MMRP19, hAPIP |
Location: |
Chr.11:34882295-34916411 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
APIP | NM_015957.3 | NP_057041.2 | ||
XM_011520154.2 | XP_011518456.1 | |||
XM_017017875.1 | XP_016873364.1 | |||
AF132963.1 | AAD27738.1 | |||
AK225249.1 | ||||
AK226157.1 | ||||
AK292648.1 | ||||
AK302225.1 | ||||
BC008440.1 | AAH08440.1 | |||
BC009077.1 | AAH09077.1 | |||
BC017594.2 | AAH17594.1 | |||
DA729016.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv832119 | Chr.11:34793981 - 34987823 on Build GRCh38 | Gain | MIR1343 APIP PDHX |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |