Assay Details
Target Gene Details
Entrez Gene ID: | 23235 |
Gene Name: | salt inducible kinase 2 |
Gene Aliases: |
LOH11CR1I, QIK, SIK-2, SNF1LK2 |
Location: |
Chr.11:111602391-111726917 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SIK2 | NM_015191.2 | NP_056006.1 | ||
XM_017017417.1 | XP_016872906.1 | |||
AB018324.3 | ||||
AB084424.1 | BAB91442.1 | |||
AB096248.1 | ||||
AK000396.1 | ||||
AK291233.1 | ||||
AL136764.2 | CAB66698.1 | |||
BC113459.1 | ||||
BC117183.1 | ||||
BM799630.1 | ||||
DA333935.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |