Assay Details
Target Gene Details
Entrez Gene ID: | 65975 |
Gene Name: | serine/threonine kinase 33 |
Gene Aliases: |
- |
Location: |
Chr.11:8335029-8594289 on Build GRCh38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
STK33 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)