Assay Details
Target Gene Details
Entrez Gene ID: | 4745 |
Gene Name: | neural EGFL like 1 |
Gene Aliases: |
IDH3GL, NRP1 |
Location: |
Chr.11:20669551-21575686 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NELL1 | NM_001288713.1 | NP_001275642.1 | ||
NM_001288714.1 | NP_001275643.1 | |||
NM_006157.4 | NP_006148.2 | |||
NM_201551.2 | NP_963845.1 | |||
AK127805.1 | ||||
AK299558.1 | ||||
AK303736.1 | ||||
AK313445.1 | ||||
BC069674.1 | AAH69674.1 | |||
BC096100.1 | AAH96100.1 | |||
BC096101.1 | AAH96101.1 | |||
BC096102.3 | AAH96102.1 | |||
BC096103.3 | AAH96103.1 | |||
BX476569.1 | ||||
D83017.1 | BAA11680.1 | |||
DA165320.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |