Assay Details
Target Gene Details
Entrez Gene ID: | 8601 |
Gene Name: | regulator of G-protein signaling 20 |
Gene Aliases: |
RGSZ1, ZGAP1, g(z)GAP, gz-GAP |
Location: |
Chr.8:53851808-53959304 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RGS20 | NM_001286673.1 | NP_001273602.1 | ||
NM_001286674.1 | NP_001273603.1 | |||
NM_001286675.1 | NP_001273604.1 | |||
NM_170587.3 | NP_733466.1 | |||
AF366054.1 | AAK54122.1 | |||
AF366055.1 | AAK54123.1 | |||
AF366056.1 | AAK54124.1 | |||
AY046538.1 | AAL03971.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)