Assay Details
Target Gene Details
Entrez Gene ID: | 9258 |
Gene Name: | malignant fibrous histiocytoma amplified sequence 1 |
Gene Aliases: |
LRRC65, MASL1 |
Location: |
Chr.8:8783354-8893621 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MFHAS1 | NM_004225.2 | NP_004216.2 | ||
AB016816.1 | BAA74737.1 | |||
AK027007.1 | ||||
BC014226.2 | AAH14226.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831228 | Chr.8:8750877 - 8933280 on Build GRCh38 | Loss |
![]() ![]() |
nsv610189 | Chr.8:8732927 - 8807430 on Build GRCh38 | Loss |
![]() |
nsv1018828 | Chr.8:8752370 - 8933150 on Build GRCh38 | Loss |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)