Assay Details
Target Gene Details
Entrez Gene ID: | 55284 |
Gene Name: | ubiquitin conjugating enzyme E2 W (putative) |
Gene Aliases: |
UBC-16, UBC16 |
Location: |
Chr.8:73780097-73878910 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
UBE2W | NM_001001481.2 | NP_001001481.2 | ||
NM_001271015.1 | NP_001257944.1 | |||
NM_018299.4 | NP_060769.4 | |||
NR_073119.1 | ||||
NR_073120.1 | ||||
NR_073121.1 | ||||
AK001873.1 | BAA91954.1 | |||
AK021735.1 | BAB13883.1 | |||
AK024050.1 | BAB14800.1 | |||
AK225554.1 | ||||
AK295792.1 | ||||
AY948289.1 | AAY24555.1 | |||
BC010900.1 | AAH10900.1 | |||
BC016326.1 | AAH16326.1 | |||
BU155172.1 | ||||
BX538145.1 | ||||
BX648427.1 | ||||
DA352184.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)