Assay Details
Target Gene Details
Entrez Gene ID: | 8767 |
Gene Name: | receptor interacting serine/threonine kinase 2 |
Gene Aliases: |
CARD3, CARDIAK, CCK, GIG30, RICK, RIP2 |
Location: |
Chr.8:89757747-89791064 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RIPK2 | NM_003821.5 | NP_003812.1 | ||
XM_005251092.3 | XP_005251149.1 | |||
AF027706.1 | AAC34970.1 | |||
AF064824.1 | AAC25668.1 | |||
AF078530.1 | AAC27722.1 | |||
AK075213.1 | ||||
AK301448.1 | ||||
AY358813.1 | AAQ89172.1 | |||
AY358814.1 | AAQ89173.1 | |||
AY524045.1 | AAS94254.1 | |||
AY562996.1 | AAS75586.1 | |||
BC004553.2 | AAH04553.1 | |||
DA040898.1 | ||||
KU178437.1 | ||||
KU178438.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3576595 | Chr.8:89459560 - 89771643 on Build GRCh38 | Gain | LOC101929709 RIPK2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |